ContactUsLinkComponent
VWR®, MOPS 10X buffer for biotechnology
VWR®, MOPS 10X buffer for biotechnology
Catalog # E526-500ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, MOPS 10X buffer for biotechnology
Catalog # E526-500ML
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    MOPS, 10X liquid concentrate, nuclease-free, makes 5 L of 1X solution
Specifications
About this item
MOPS is a zwitterionic buffer used as a running buffer for denaturing agarose gel electrophoresis of RNA. Having a pH buffering range from 6,5 to 7,9, MOPS works exceptionally well with formaldehyde gels at 20 mM concentration.
  • Optimised for RNA agarose electrophoresis
  • Ideal for use with RNA formaldehyde gels