Order Entry
Austria
ContactUsLinkComponent
cis-Permethrin
cis-Permethrin
Catalog # EHERC15990100
Supplier:  EHRENSTORFER
CAS Number:  
cis-Permethrin
Catalog # EHERC15990100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
EINECS:263-346-3
CAS Number:61949-76-6
Formula:C₂₁H₂₀Cl₂O₃