Order Entry
Austria
Orders LinkContactUsLinkComponent
Ectoine

Ectoine

Ectoine
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  (S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
  • CAS Number:96702-03-3
  • Formula:C₆H₁₀N₂O₂
  • MDL Number:MFCD03419286
  • Molecular Weight:142.1558 g/mol
  • EINECS:431-910-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g