Order Entry
Austria
ContactUsLinkComponent
Phenylmagnesium bromide 3 mol/l
Phenylmagnesium bromide 3 mol/l
Catalog # H54729.AE
CAS Number:  
Phenylmagnesium bromide 3 mol/l
Catalog # H54729.AE
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

About this item

 :
 :181.31 g/mol
Storage Temperature:Ambient
EINECS:202-867-2
UN:2924
ADR:3,II
 :GHS02
 :GHS05
CAS Number:100-58-3
Formula:C₆H₅BrMg
MDL Number:MFCD00000038