Order Entry
Austria
ContactUsLinkComponent
Herbicide reference standard, Dimethenamid-P 10 µg/ml in methanol, Dr Ehrenstorfer
Herbicide reference standard, Dimethenamid-P 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL12678000ME
Supplier:  EHRENSTORFER
CAS Number:  
Herbicide reference standard, Dimethenamid-P 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL12678000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    163515-14-8
  • Description:
    Dimethenamid-P 10 µg/ml in methanol
  • Formula:
    C₁₂H₁₈ClNO₂S
  • Description:
    Dimethenamid-P

Specifications

About this item