Pesticide reference standard, Bifenthrin 100 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERXA10584000IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:82657-04-3
- Description:Bifenthrin 100 µg/ml in isooctane
- Formula:C₂₃H₂₂ClF₃O₂
- Description:Bifenthrin