Pesticide reference standard, cis-Permethrin 100 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERXA15990100AL
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:61949-76-6
- Description:cis-Permethrin 100 µg/ml in acetonitrile
- Formula:C₂₁H₂₀C₁₂O₃
- Description:cis-Permethri