Phosphamidon (mixture of cis and trans isomers), Supelco®
Catalog # 45622-100MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:236-116-5
UN:2810
CAS Number:13171-21-6
Formula:C₁₀H₁₉ClNO₅P