Order Entry
Austria
Orders LinkContactUsLinkComponent
 

β-Cyclodextrine

Glass bottle, 5 mL
Catalog # BECL713331
Marketsource
Supplier:   Beckman Coulter
β-Cyclodextrine
Catalog # BECL713331
Supplier:  Beckman Coulter
Marketsource
CAS Number:  7585-39-9
β-Cyclodextrine

Glass bottle, 5 mL

CAS Number:  7585-39-9
Price
Inquire For Price
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • EINECS:231-493-2
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.