Order Entry
Austria
Orders LinkContactUsLinkComponent
β-Cyclodextrine

β-Cyclodextrine

Glass bottle for solids, 100 g
Catalog # A2147.0100
Supplier:   PanReac AppliChem
β-Cyclodextrine
Catalog # A2147.0100
Supplier:  PanReac AppliChem
CAS Number:  7585-39-9
β-Cyclodextrine

Glass bottle for solids, 100 g

CAS Number:  7585-39-9
Price
210,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • EINECS:231-493-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.