Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Iodo-5-nitropyridine
2-Iodo-5-nitropyridine, Sigma-Aldrich®
2-Iodo-5-nitropyridine, Sigma-Aldrich®
SIAL647004-1G
:
Merck
:
2-Iodo-5-nitropyridine, Sigma-Aldrich®
SIAL647004-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
249.98 g/mol
:
M-Clarity™ quality level = MQ100
:
28080-54-8
:
C₅H₃IN₂O₂
:
MFCD05863241