Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Alachlor
Alachlor
Alachlor
Alachlor
Catalog # EHERC10060000
Supplier:
EHRENSTORFER
CAS Number:
Certificates
Expand image
Alachlor
Catalog # EHERC10060000
Supplier:
EHRENSTORFER
CAS Number:
Certificates
Specifications
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
Specifications
About this item
Product Details
Documentation
:
EINECS:
240-110-8
CAS Number:
15972-60-8
Formula:
C₁₄H₂₀ClNO₂
Product Details
Documentation