Pesticide reference standard, Methomyl (1-(Methylthio)ethylideneamino N-methylcarbamate) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERXA15030000CY
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:16752-77-5
- Description:Methomyl (1-(Methylthio)ethylideneamino N-methylcarbamate) 100 µg/ml in cyclohexane
- Formula:C₅H₁₀N₂O₂S
- Description:Methomy