Order Entry
Austria
ContactUsLinkComponent
 
Pesticide reference standard, Methomyl (1-(Methylthio)ethylideneamino N-methylcarbamate) 100 µg/ml in acetonitrile
  PROONE7251A
 :  
Pesticide reference standard, Methomyl (1-(Methylthio)ethylideneamino N-methylcarbamate) 100 µg/ml in acetonitrile
  PROONE7251A
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    16752-77-5
  • Description:
    Methomyl (1-(Methylthio)ethylideneamino N-methylcarbamate) 100 µg/ml in acetonitrile
  • Formula:
    C₅H₁₀N₂O₂S
  • Description:
    Methomy