Order Entry
Austria
ContactUsLinkComponent
Dipropyleneglycol monomethyl ether (mixture of isomers)
Dipropyleneglycol monomethyl ether (mixture of isomers)
Catalog # KAMM2035495
Supplier:  KAMMERER
CAS Number:  
Dipropyleneglycol monomethyl ether (mixture of isomers)
Catalog # KAMM2035495
Supplier:  KAMMERER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :148.2001 g/mol
EINECS:252-104-2
CAS Number:34590-94-8
Formula:C₇H₁₆O₃
MDL Number:MFCD00059604