Order Entry
Austria
ContactUsLinkComponent
Pesticide reference standard, Vamidothion 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Pesticide reference standard, Vamidothion 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA17900000AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Vamidothion 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA17900000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    2275-23-2
  • Description:
    Vamidothion 100 µg/ml in acetonitrile
  • Formula:
    C₈H₁₈NO₄PS₂
  • Description:
    Vamidothion

Specifications

About this item