2-Dimethylaminoethyl methacrylate stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2867-47-2
:
Formula:C₈H₁₅NO₂
MDL Number:MFCD00008589
:157.21 g/mol
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
EINECS:220-688-8
UN:2522
Frequently Bought Together