2-Dimethylaminoethyl methacrylate stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2867-47-2
:
Formula:C₈H₁₅NO₂
MDL Number:MFCD00008589
:157.21 g/mol
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
EINECS:220-688-8
UN:2522
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-50 °C
- Molar Mass:157.21 g/mol
- Boiling Point:182 - 192 °C (1013 hPa)
- Vapor Pressure:1 hPa (20 °C)
- Flash Point:65 °C
- Explosion Limit:1.20 %(V)
- Density:0.93 g/cm3 (20 °C)
- Ignition Point:255 °C
Specification Test Results
Frequently Bought Together