Monuron (N-(4-chlorophenyl)-N',N'-dimethylurea)
Catalog # EHERC15320000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:198.65 g/mol
EINECS:205-766-1
CAS Number:150-68-5
Formula:C₉H₁₁ClN₂O
MDL Number:MFCD00018556