Order Entry
Austria
ContactUsLinkComponent
Glycine-tert-butyl ester(H-Gly-OtBu) for synthesis, Sigma-Aldrich®
Glycine-tert-butyl ester(H-Gly-OtBu) for synthesis, Sigma-Aldrich®
Catalog # 8.18213.0006
Supplier:  Merck
CAS Number:  
Glycine-tert-butyl ester(H-Gly-OtBu) for synthesis, Sigma-Aldrich®
Catalog # 8.18213.0006
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 ml
  • Seq:
    2210

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    131.17 g/mol
  • Boiling Point:
    29 - 31 °C (3 hPa)
  • Refractive Index:
    1.4237 (20 °C, 589 nm)
  • Density:
    0.96 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :131.18 g/mol
 :M-Clarity™ quality level = MQ200
CAS Number:6456-74-2
Formula:C₆H₁₃NO₂
MDL Number:MFCD00038194