ContactUsLinkComponent
Food butyrometer, without stopper
Food butyrometer, without stopper
Catalog # 614-0135
Supplier:  GERBER FUNKE
Food butyrometer, without stopper
Catalog # 614-0135
Supplier:  GERBER FUNKE
Specifications
  • Description:
    Butyrometer for food, Roeder´s, 0 - 100 % : 1,0
  • Pk:
    10
Specifications
About this item
According to Roeder´s weighing method.