About this item
Spring/lock Stainless steel V4A
Fast and safe connection for liquids, gases, compressed air and vacuum. Rapid couplings are the ideal when connecting and separating hoses, machines, apparatus, containers, etc. Top quality, with tested reliability, small space requirement, maximum possible flow, easy operation (single handed operation) and an elegant design distinguish the rapid couplings.The couplings are available in two different materials: POM (Polyoxymethylene) and PP (Polypropylene). The rapid couplings are available with a hose nozzle, hose screw joint, thread connector or as a built-in component for control panels, with or without stop-valve respectively.The couplings are simply connected to each other, automatically clicking intoplace and opening the valves.When disconnecting, (one handed operation), the valves are automatically sealed, no leakage occurs.
POM
Seal (standard): Buna-N
Operating pressure at 20 °C with vacuum up to 10 bar
Operating temperature: −40 to +80 °C
PP
Seal (standard): EPDM
Operating pressure at 20 °C: up to 8 bar
Operating temperature: 0 to +110 °C