Order Entry
Austria
Orders LinkContactUsLinkComponent
Urine specific refractometer, UG-α

Urine specific refractometer, UG-α

Urine specific refractometer, UG-α, 1,00 − 1,06 °Bx
Catalog # ATAG3464
Supplier:   Atago
Urine specific refractometer, UG-α
Catalog # ATAG3464
Supplier:  Atago
Urine specific refractometer, UG-α
Urine specific refractometer, UG-α

Urine specific refractometer, UG-α, 1,00 − 1,06 °Bx

Price
989,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This refractometer is designed to use the index method for measuring total protein in serum.

  • Streaming digital LCD display of urine specific gravity
  • Automatic temperature compensation (ATC) for immediate, accurate calibration readings, regardless of ambient temperature
  • External light interference (ELI) feature displays warning message when direct light is detected

The protein scale has been prepared after extensive clinical analyses.

Delivery information: Supplied with a battery, a protective case and instructions.

Specifications
  • Model:
    UG-α
  • Range:
    1,00 − 1,06 °Bx
  • Description:
    Urine specific refractometer
  • Cat. No.:
    ATAG3464
  • Accuracy:
    ±0,001 °Bx
  • Sample size:
    0,1 ml
  • W×D×H:
    170×90×40 mm
  • Electrical:
    9 V dry battery
  • Temperature compensation range:
    10…35 °C
  • Weight:
    300 g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.