Centrifuge tubes, conical bottom
: KARTELL
PP, translucent.
- Very good chemical resistance
- Very good temperature resistance
- Graduated
Conical bottom with tight screw cap.
Frequently Bought Together
PP, translucent.
Conical bottom with tight screw cap.