Foam head swabs
Supplier: HERENZ HEINZ
About this item
Swabs with PU foam heads and plastic applicators.
- Fuzzfree
Designed for bacterial sampling or computer cleaning.
2 Options Available Below
Product Details & Documents
Swabs with PU foam heads and plastic applicators.
- Fuzzfree
Designed for bacterial sampling or computer cleaning.
Specifications for Product Family
Specifications
- Catalog No:
- HERE1020009
- HERE1020011
- Total length:
- 150,0 mm
- 130,0 mm
Specifications
Product Family Options
Product Information
- Shaft/headLengthAvailabilityPrice
- HERE1020009Swabs with big foam headSpecifications:
More
Specifications
Cat. No.HERE1020009Total length150,0 mm - HERE1020011Swabs with flexible foam headSpecifications:
More
Specifications
Cat. No.HERE1020011Total length130,0 mm