Handheld refractometers, 'PEN' series
: Atago
Designed for continuous measurement to expand your current methods.
- Compact design for one-handed operation
- Automatic temperature compensation (ATC)
- Rinse off the sample by running water on the prism
: IP 65 to rinse off the sample with running water
: Supplied in a storage case with battery.
- Catalog No:
- ATAG3741
- ATAG3730
- Model:
- PEN-Urine S.G.
- PEN-PRO
- Response time:
- 2 seconds
- 2 seconds
- Weight:
- 70 g
- 70 g
Frequently Bought Together