illustra™ MicroSpin™ S-300 HR columns
Artikelnr. 27-5130-01
Supplier: Cytiva
CAS Number:
Specifications
- Pk:50
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:Illustra™ MicroSpin™ S-300 HR columns
- Description:Illustra™ MicroSpin™ S-300 HR columns
Specifications
About this item
illustra™ MicroSpin™ S-300 HR columns designed for rapid purification of PCR products (>100 bp) from unincorporated primers (<20-mers) and nucleotides using spin-column chromatography.
- Accommodates 25 to 50 μl for post-PCR clean-up prior to sequencing
- Useful for purification of alkaline-denatured plasmid DNA prior to sequencing
- Prepacked for convenience with Sephacryl™ S-300 HR pre-equilibrated in TE buffer
- Ready to use, requiring less than 4 minutes from sample application to collection of purified product
- Tested in nickase, single- and double-stranded exonuclease and RNase assays