Orders LinkContactUsLinkComponent
Chairs, Fabric
Supplier:   Kango
Chairs, Fabric
Supplier:  Kango
Certificates
Chairs, Fabric
16 Options Available Below
Product Details & Documents
Chairs with high density comfortable foam and fabric coating
  • Ergonomically designed seat
  • Available with castors or glides
  • Can also be equipped with a footring
  • Gas lift column with height adjustment and shok absorber
  • 5-branch scuff resistant reinforced base
Chairs:
Shaped backrest with rounded edges
Articulated backrest, adjustable in height and depth (adjustable chairs)Independently adjustable seat and backrest tilt with one fingertip lever thanks to an asynchronous mechanism (for asynchronous chairs)Rectangular seat
Dimensions of the seat (W×D): 435×390 mm
Dimensions of the backrest (W×H): 410×250 mm
Product Family Options
Adjustable chairs
Product Information
  • Height
    Foot style
    Foot ring
    With
    Availability
    Price
  • 132-8434
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    400 - 530 mm
    Glides
    Chrome gas-lift column, with height adjustment and shock absorber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 275,00 €
    275,00 €
    1 * 1 ST
       275,00 €
  • BIEN6NN40GBLP00705
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    410 - 540 mm
    Glides
    Chrome gas-lift column, with height adjustment
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 328,00 €
    328,00 €
    1 * 1 ST
       328,00 €
  • BIEN6NN35NBPP00705
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    430 - 560 mm
    Glides
    Black gas-lift column, with height adjustment and shock absorber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 247,00 €
    247,00 €
    1 * 1 ST
       247,00 €
  • 132-8437
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    440 - 570 mm
    Castors
    Chrome gas-lift column, with height adjustment and shock absorber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 279,00 €
    279,00 €
    1 * 1 ST
       279,00 €
  • BIEN6NN40GBLR00705
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    450 - 580 mm
    Castors
    Chrome gas-lift column, with height adjustment
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 333,00 €
    333,00 €
    1 * 1 ST
       333,00 €
  • BIEN6NN35NBPR00705
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    470 - 600 mm
    Castors
    Black gas-lift column, with height adjustment and shock absorber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 252,00 €
    252,00 €
    1 * 1 ST
       252,00 €
  • 132-8096
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    510 - 640 mm
    Castors
    Chrome gas-lift column, with height adjustment and shock absorber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 294,00 €
    294,00 €
    1 * 1 ST
       294,00 €
  • 132-8095
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    510 - 640 mm
    Castors
    Height adjustable, PA
    Chrome gas-lift column, with height adjustment and shock absorber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 338,00 €
    338,00 €
    1 * 1 ST
       338,00 €
  • 132-7894
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    540 - 780 mm
    Glides
    Chrome gas-lift column, with height adjustment and shock absorber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 286,00 €
    286,00 €
    1 * 1 ST
       286,00 €
  • 132-8435
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    540 - 780 mm
    Glides
    Height adjustable, PA
    Chrome gas-lift column, with height adjustment and shock absorber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 331,00 €
    331,00 €
    1 * 1 ST
       331,00 €
  • BIEN6NN40GHLP00705
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    550 - 790 mm
    Glides
    Chrome gas-lift column, with height adjustment
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 341,00 €
    341,00 €
    1 * 1 ST
       341,00 €
  • BIEN6NN40GHLP01705
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    550 - 790 mm
    Glides
    Height adjustable, aluminium
    Chrome gas-lift column, with height adjustment
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 396,00 €
    396,00 €
    1 * 1 ST
       396,00 €
  • BIEN6NN35NHPP00705
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    570 - 810 mm
    Glides, articulated
    Black gas-lift column, with height adjustment
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 259,00 €
    259,00 €
    1 * 1 ST
       259,00 €
  • BIEN6NN35NHPP02705
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    Ergonomic seat in grey fabric, with articulated backrest, adjustable in depth and height , with polyamide base
    570 - 810 mm
    Glides, articulated
    Height adjustable, PA
    Black gas-lift column, with height adjustment
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 307,00 €
    307,00 €
    1 * 1 ST
       307,00 €
  • BIEN6NN35GHLR02705
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    580 - 820 mm
    Castors
    Height adjustable, PA
    Chrome gas-lift column, with height adjustment and shock absorber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 335,00 €
    335,00 €
    1 * 1 ST
       335,00 €
  • BIEN6NN35GTLP01705
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    Ergonomic seat in grey fabric, scuff resistant, 5-branch and reinforced base, with articulated backrest, adjustable in depth and height
    720 - 960 mm
    Glides
    Height adjustable, aluminium
    Chrome gas-lift column, with height adjustment and shock absorber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 356,00 €
    356,00 €
    1 * 1 ST
       356,00 €