ContactUsLinkComponent
Pipette tips, Matrix®
Pipette tips, Matrix®
  613-1836
Pipette tips, Matrix®
  613-1836
  • Packed:
    10 racks, 96 each (Ecotips refill)
  • Sterile:
    -
  • Volume:
    1250 µl
  • Seq:
    35
  • Tip style:
    Extended
  • Pk:
    960
Transparent PP, sterile or non sterile.
  • Sealing rings on tips ensure confident, leak-free seal on most pipettes
  • Tip manufactured under tight QC specifications ensures low liquid retention without special treatments or additives
  • Extended tips allow access to the bottom of test tubes, reagent bottles, flasks and other vessels without touching the pipette barrel against the side of the tube
 : Available bulk, in Flo-Thru racks or in Ecotips refills. Flo-Thru racks individually support tips to prevent them from wobbling when trying to seat them on the pipette. Ecotips refill system is 100% recyclable and offers fast rack reloading in a space saving design.