ContactUsLinkComponent
Griffin beakers, PP
Griffin beakers, PP
Catalog # BURK7122-0500
Supplier:  BURKLE
Supplier #: 7122-0500
undefined
Griffin beakers, PP
Catalog # BURK7122-0500
Supplier:  BURKLE
Supplier #: 7122-0500
Log in for availability
Select an Option
94 mm
Select an Option
50 ml
Select an Option
500 ml
Select an Option
118 mm
Select an Option
05
Select an Option
1
About this item
Laboratory beakers according to DIN 12331 and ISO 7056 (Glass) for various uses.
  • PP, highly transparent
  • Blue graduation printed on
  • Shatterproof
When bought in bulk, individual print is possible.
Certifications: ISO 7056
Frequently Bought Together