ContactUsLinkComponent
SP Bel-Art® precisionware sampler for acids
SP Bel-Art® precisionware sampler for acids
Catalog # BELAH378010000
undefined
SP Bel-Art® precisionware sampler for acids
Catalog # BELAH378010000
Specifications
  • Capacity:
    500 ml
  • Colour:
    White
  • Description:
    Sampler for chemicals
  • Length:
    142 cm
  • Material:
    PP
  • Pk:
    1
Specifications
About this item
PE bottle threads on to end of rod to collect samples. Safely distances user from dangerous chemicals.
  • Fluid enters through fitting above bottle
  • T-handle for secure grip when lowering sampler
  • Handle has two piece threaded joint to shorten during transport or storage
Designed for hydrochloric, sulfuric and acetic acids, formaldehyde potassium or sodium hydroxide, in tanks rail cars, and other large vessels.