ContactUsLinkComponent
Scriber pen, Glascribe®, SP Bel-Art
  811-0103
undefined
Scriber pen, Glascribe®, SP Bel-Art
  811-0103
  • Description:
    Marker pen, with retractable tungsten carbide tip
  • Pk:
    1
The Glascribe® Pen, with retractable tungsten carbide tip, lets you inscribe on glass, ceramics, and plastic.
  • With pocket clip, so it is always handy when needed