Orders LinkContactUsLinkComponent
Face shields
Face shields
Supplier:   UVEX
Face shields
Supplier:  UVEX
Certificates
Face shields
2 Options Available Below
Product Details & Documents
Robust visor with integrated forehead cover, clear cellulose acetate lens and additional protective cap made from impact-resistant PS.
  • Twist ratchet enables simple and reliable adjustment to any head size
  • Flip-up mechanism allows the visor to be set in any position
  • Easy lens replacement
Certifications: EN 166
Documents
Product Family Options
Product Information
  • Type
    Material
    L×W
    Availability
    Price
  • Anti-UV (UV 380)
    Cellulose acetate
    195×412 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,90 €
    40,90 €
    1 * 1 ST
       40,90 €
  • Anti-UV (UV 380), anti-fog
    Cellulose acetate
    195×540 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,80 €
    43,80 €
    1 * 1 ST
       43,80 €
Accessories
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor lens
    Replacement visor lens
    111-2543
    Cellulose acetate
    Log in for availability
    1 * 1 ST - 13,70 €
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor lens, anti-fog
    Replacement visor lens, anti-fog
    111-8229
    Cellulose acetate
    Log in for availability
    1 * 1 ST - 30,70 €