ContactUsLinkComponent
Forceps, SP Bel-Art
Forceps, SP Bel-Art
  BELAH379370000
undefined
Forceps, SP Bel-Art
  BELAH379370000
  • Type:
    Fully coated
  • Length:
    98 mm
  • Seq:
    03
  • Pk:
    2
Stainless steel, Teflon® FEP coated (partly of fully). The brown Teflon® FEP coating on these stainless steel forceps will not scratch or mar delicate surfaces, while its anti-stick qualities prevent material build-up.
  • Coating complies with FDA regulations for direct contact with food
  • All forceps are 98 mm (3⅞") long with medium points and serrated tips