ContactUsLinkComponent
Forceps, curved, SP Bel-Art
Forceps, curved, SP Bel-Art
  BELAH379400000
undefined
Forceps, curved, SP Bel-Art
  BELAH379400000
  • Type:
    Curved, serrated
  • Length:
    98 mm
  • Pk:
    2
Gently curved stainless steel forceps with exceptionally fine tips which are coated with brown Fluo-Kem® PTFE.
  • FDA compliant for food contact
  • Serrated, curved tips