Stirrer blades for centrifugal stirrer shafts, BOLA
Supplier: Bohlender
Certificates
About this item
Centrifugal stirrer blades made of PTFE consisting of 2 paddles, bolt and clamp ring.
- Resistant to chemicals
- Temperature resistance from –200 to +250 °C
Suitable for vessels with a 24/29 ground joint and centrifugal stirrer shafts.
Frequently Bought Together