Orders LinkContactUsLinkComponent
Tempering beakers
Tempering beakers
Supplier:   KGW
Tempering beakers
Supplier:  KGW
Certificates
Tempering beakers
Tempering beakers
4 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Coolant ram pressure max. 0,5 bar
  • High thermal resistance
  • Very good chemical resistance
For tempering of liquids. The tempering medium is pumped round the double walled casing with the aid of a thermostat.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 250 ml
    120 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
  • 600 ml
    150 mm
    95 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 182,00 €
    182,00 €
    1 * 1 ST
       182,00 €
  • 2000 ml
    210 mm
    135 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 377,00 €
    377,00 €
    1 * 1 ST
       377,00 €
  • 600 ml
    150 mm
    95 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €