Stirrer blades, moon-shaped, with angular groove
Supplier: Bohlender
Certificates
About this item
Stirrer blades made of PTFE, with angular groove and 5,8 mm bore diameter.
- Resistant to chemicals
- Temperature resistance from –200 to +250 °C
Suitable for vessels with a 29/32 ground joint and glass stirrer shafts and KPG glass stirrer shafts.
Frequently Bought Together