Order Entry
Austria
ContactUsLinkComponent
UV Incubator, benchtop, SI-950
UV Incubator, benchtop, SI-950
Catalog # UVPA95-0358-02
undefined
UV Incubator, benchtop, SI-950
Catalog # UVPA95-0358-02

Specifications

  • W×D×H ext.:
    445×375×457 mm
  • Capacity:
    26,9 L
  • Temperature range:
    max. 68 °C
  • Description:
    Benchtop incubator with built-in shortwave UV and two ventilated shelves
  • Model:
    SI-950
  • Ext. W×D×H:
    445×375×457 mm
  • Shelves supplied (max.):
    Two ventilated shelves
  • Pk:
    1

Specifications

About this item

This benchtop UV Incubator with stainless steel interior is designed to provide precise temperature control and uniformity throughout the chamber. Used for incubation of biological assays, fungal, bacterial cultures, eggs and other biological samples.

  • Microprocessor controlled temperature
  • View samples without opening the door
  • Temperature: 68 °C maximum
  • Shelves: adjustable at three positions
  • Overhead shortwave 254 nm ultraviolet for sterilisating the chamber between experiments
Delivery: Supplied with two ventilated shelves.