Powder funnels, SP Bel-Art
PP, translucent.
- Very good chemical resistance
- Very good temperature resistance
- Autoclavable
Specially designed for transferring powders. These funnels have 60º angle and a wide stem for clog-free filling.
Frequently Bought Together