Order Entry
Austria
ContactUsLinkComponent
Vanadium standard solution, concentrate, 1000 mg V in 8,6% sulphuric acid (from VO(SO₄)) Titrisol® standard for AAS, Supelco®
Catalog # 1.09994.0001
Supplier:  Merck
CAS Number:  
Vanadium standard solution, concentrate, 1000 mg V in 8,6% sulphuric acid (from VO(SO₄)) Titrisol® standard for AAS, Supelco®
Catalog # 1.09994.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg
  • Matrix:
    8,6% H₂SO₄
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Vanadium
  • Symbol:
    V

Specifications

About this item

Certifications: Traceable to SRM from NIST