Order Entry
Austria
ContactUsLinkComponent
 
Titanium standard solution, concentrate, 1000 Ti in water (from (NH₄)₂TiF₆) Titrisol® standard for AAS, Supelco®
  1.09829.0001
 :  Merck
 :  
Titanium standard solution, concentrate, 1000 Ti in water (from (NH₄)₂TiF₆) Titrisol® standard for AAS, Supelco®
  1.09829.0001
 :  Merck
 :  

 

  • Concentration:
    1000 mg
  • Matrix:
    H₂O
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Element:
    Titanium
  • Symbol:
    Ti

 

 

 : Traceable to SRM from NIST