Order Entry
Austria
ContactUsLinkComponent
Silver standard solution, concentrate, 1000 mg Ag in 5% nitric acid (from AgNO₃) Titrisol® standard for AAS, Supelco®
Silver standard solution, concentrate, 1000 mg Ag in 5% nitric acid (from AgNO₃) Titrisol® standard for AAS, Supelco®
Catalog # 1.09906.0001
Supplier:  Merck
CAS Number:  
Silver standard solution, concentrate, 1000 mg Ag in 5% nitric acid (from AgNO₃) Titrisol® standard for AAS, Supelco®
Catalog # 1.09906.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Silver
  • Symbol:
    Ag

Specifications

About this item

Certifications: Traceable to SRM from NIST