Order Entry
Austria
ContactUsLinkComponent
 
Molybdenum standard solution, concentrate, 1000 Mo Mn in water with 0,7% ammonium hydroxide (from (NH₄)₆Mo₇O₂₄) Titrisol® standard for AAS, Supelco®
  1.09926.0001
 :  Merck
 :  
Molybdenum standard solution, concentrate, 1000 Mo Mn in water with 0,7% ammonium hydroxide (from (NH₄)₆Mo₇O₂₄) Titrisol® standard for AAS, Supelco®
  1.09926.0001
 :  Merck
 :  

 

  • Concentration:
    1000 mg
  • Matrix:
    H₂O/tr.NH₄OH
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Molybdenum
  • Symbol:
    Mo

 

 

 : Traceable to SRM from NIST