Order Entry
Austria
ContactUsLinkComponent
 
Magnesium standard solution, concentrate, 1000 mg Mg in 6% hydrochloric acid (from MgCl₂) Titrisol® standard for AAS, Supelco®
Catalog # 1.09949.0001
Supplier:  Merck
CAS Number:  
Magnesium standard solution, concentrate, 1000 mg Mg in 6% hydrochloric acid (from MgCl₂) Titrisol® standard for AAS, Supelco®
Catalog # 1.09949.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg
  • Matrix:
    6% HCl
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item

Certifications: Traceable to SRM from NIST