Potassium standard solution, concentrate, 1000 mg K in water (from KCl) Titrisol® standard for AAS, Supelco®
Artikelnr. 1.09924.0001
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg
- Matrix:H₂O
- Pack type:Plastic ampoule
- Pk:1 Ampoul
- M-Clarity quality level:MQ100
- Element:Potassium
- Symbol:K
Specifications
About this item
Certifications: Traceable to SRM from NIST