Order Entry
Austria
ContactUsLinkComponent
Chromium standard solution, concentrate, 1000 mg Cr in 4,2% hydrochloric acid (from CrCl₃) Titrisol® standard for AAS, Supelco®
Chromium standard solution, concentrate, 1000 mg Cr in 4,2% hydrochloric acid (from CrCl₃) Titrisol® standard for AAS, Supelco®
Catalog # 1.09948.0001
Supplier:  Merck
CAS Number:  
Chromium standard solution, concentrate, 1000 mg Cr in 4,2% hydrochloric acid (from CrCl₃) Titrisol® standard for AAS, Supelco®
Catalog # 1.09948.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HCl
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Chromium
  • Symbol:
    Cr

Specifications

About this item

Certifications: Traceable to SRM from NIST