Order Entry
Austria
ContactUsLinkComponent
Calcium standard solution, concentrate, 1000 mg Ca in 6,5% hydrochloric acid (from CaCl₂) Titrisol® standard for AAS, Supelco®
Calcium standard solution, concentrate, 1000 mg Ca in 6,5% hydrochloric acid (from CaCl₂) Titrisol® standard for AAS, Supelco®
Catalog # 1.09943.0001
Supplier:  Merck
CAS Number:  
Calcium standard solution, concentrate, 1000 mg Ca in 6,5% hydrochloric acid (from CaCl₂) Titrisol® standard for AAS, Supelco®
Catalog # 1.09943.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HCl
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item

CaCl₂ in 6,5% hydrochloric acid

Certifications: Traceable to SRM from NIST