Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-(-)-1,1'-Bi-2-naphthol
(S)-(-)-1,1'-Bi-2-naphthol
(S)-(-)-1,1'-Bi-2-naphthol
MOLE11057305-1G
:
Molekula
:
(S)-(-)-1,1'-Bi-2-naphthol
MOLE11057305-1G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
286.33 g/mol
:
2811
:
6.1,III
:
18531-99-2
:
C₂₀H₁₄O₂
:
MFCD00004068