Order Entry
Austria
ContactUsLinkComponent
Certipur®, Supelco® Thorium Standard Solution, 10 mg/L Th in dil. Nitric Acid (from Th(NO₃)₄), Standard for ICP
Certipur®, Supelco® Thorium Standard Solution, 10 mg/L Th in dil. Nitric Acid (from Th(NO₃)₄), Standard for ICP
Catalog # 1.70391.0100
Supplier:  Merck
CAS Number:  
undefined
Certipur®, Supelco® Thorium Standard Solution, 10 mg/L Th in dil. Nitric Acid (from Th(NO₃)₄), Standard for ICP
Catalog # 1.70391.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • GHS signal:
    Warning
  • Storage temperature:
    Ambient
  • Element:
    Thorium
  • Symbol:
    Th

Specifications

About this item

Certifications: Traceable to SRM from NIST.